Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234570.2 | XP_006234632.1 | 2 - 3 | 265 | 59 | |
GenBank mRNA | BC167758.1 | - | 2 - 3 | 214 | 7123311 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |