Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hivep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107972.1 | NP_001101442.1 | 3 - 4 | 5761 | 69 | |
XM_006238779.2 | XP_006238841.1 | 4 - 5 | 8913 | 69 | ||
XM_008764014.1 | XP_008762236.1 | 3 - 4 | 9010 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |