Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037556.2 | NP_001032645.2 | 2 - 3 | 210 | 68 | |
XM_006239429.2 | XP_006239491.1 | 2 - 3 | 844 | 68 | ||
XM_006239430.2 | XP_006239492.1 | 2 - 3 | 429 | 68 | ||
XM_006239431.2 | XP_006239493.1 | 1 - 2 | 171 | 68 | ||
XM_008764237.1 | XP_008762459.1 | 3 - 4 | 367 | 68 | ||
XM_008764238.1 | XP_008762460.1 | 2 - 3 | 192 | 68 | ||
GenBank mRNA | AY788917.1 | - | 2 - 3 | 204 | 68 | |
BC127446.1 | - | 2 - 3 | 210 | 7383226 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |