Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115034.1 | NP_001108506.1 | 5 - 6 | 697 | 89 | |
GenBank mRNA | BC158698.1 | - | 5 - 6 | 654 | 9027714 | 89 |
FQ223544.1 | - | 5 - 6 | 681 | 89 | ||
FQ232113.1 | - | 5 - 6 | 659 | 89 | ||
FQ233084.1 | - | 5 - 6 | 659 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |