Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109445.1 | NP_001102915.1 | 8 - 9 | 648 | 75 | |
XM_006240399.2 | XP_006240461.1 | 8 - 9 | 622 | 75 | ||
GenBank mRNA | BC158833.1 | - | 8 - 9 | 590 | 7381517 | 75 |
FJ179410.1 | - | 7 - 8 | 525 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |