Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108047.1 | NP_001101517.1 | 4 - 5 | 875 | 62 | |
XM_006240429.2 | XP_006240491.1 | 2 - 3 | 673 | 62 | ||
XM_006240431.2 | XP_006240493.1 | 3 - 4 | 1511 | 62 | ||
XM_008764800.1 | XP_008763022.1 | 3 - 4 | 2248 | 62 | ||
XM_008764801.1 | XP_008763023.1 | 4 - 5 | 943 | 62 | ||
XM_008764802.1 | XP_008763024.1 | 3 - 4 | 2248 | 62 | ||
XM_008764803.1 | XP_008763025.1 | 3 - 4 | 2248 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |