Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107718.2 | NP_001101188.2 | 5 - 6 | 1012 | 65 | |
NM_001134856.1 | NP_001128328.1 | 5 - 6 | 938 | 65 | ||
XM_008761460.1 | XP_008759682.1 | 6 - 7 | 462 | 65 | ||
GenBank mRNA | BC161876.1 | - | 5 - 6 | 1012 | 7115002 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |