Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191984.1 | NP_001178913.1 | 3 - 4 | 450 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |