Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034922.1 | NP_001030094.1 | 2 - 3 | 328 | 65 | |
GenBank mRNA | BC105762.1 | - | 2 - 3 | 328 | 7929859 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |