Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109271.1 | NP_001102741.1 | 6 - 7 | 690 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |