Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017227.1 | NP_058923.1 | 12 - 13 | 1469 | 93 | |
GenBank mRNA | AB008803.1 | - | 12 - 13 | 1469 | 93 | |
AB010999.1 | - | 12 - 13 | 1469 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |