Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130405.1 | NP_569089.1 | 2 - 3 | 591 | 98 | |
GenBank mRNA | AF305619.1 | - | 2 - 3 | 591 | 98 | |
AF393783.1 | - | 2 - 3 | 508 | 98 | ||
BC061987.1 | - | 2 - 3 | 615 | 5599491 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |