Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006979.1 | NP_001006980.1 | 7 - 8 | 1474 | 63 | |
GenBank mRNA | BC083869.1 | - | 7 - 8 | 1474 | 7128388 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |