Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751737.3 | XP_003751785.1 | 2 - 3 | 460 | 96 | |
XM_008771924.1 | XP_008770146.1 | 2 - 3 | 392 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |