Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107450.1 | NP_001100920.1 | 4 - 5 | 780 | 149 | |
XM_008761497.1 | XP_008759719.1 | 4 - 5 | 1932 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |