Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022297.2 | NP_071633.1 | 3 - 4 | 584 | 95 | |
GenBank mRNA | FQ220049.1 | - | 3 - 4 | 604 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |