Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138851.1 | NP_620206.1 | 1 - 2 | 72 | 65 | |
GenBank mRNA | AY089983.1 | - | 1 - 2 | 72 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |