Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013913.1 | NP_001013935.1 | 3 - 4 | 397 | 122 | |
XM_008761384.1 | XP_008759606.1 | 3 - 4 | 568 | 122 | ||
XM_008761385.1 | XP_008759607.1 | 3 - 4 | 571 | 122 | ||
XM_008761386.1 | XP_008759608.1 | 3 - 4 | 566 | 122 | ||
XM_008761387.1 | XP_008759609.1 | 3 - 4 | 566 | 122 | ||
XM_008761388.1 | XP_008759610.1 | 3 - 4 | 572 | 122 | ||
XM_008761389.1 | XP_008759611.1 | 3 - 4 | 560 | 122 | ||
XM_008761390.1 | XP_008759612.1 | 3 - 4 | 561 | 122 | ||
GenBank mRNA | BC085920.1 | - | 3 - 4 | 397 | 7108742 | 122 |
FQ209547.1 | - | 3 - 4 | 400 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |