Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016986.2 | NP_058682.2 | 4 - 5 | 394 | 93 | |
GenBank mRNA | CK359511.1 | - | 4 - 5 | 309 | 93 | |
J02791.1 | - | 4 - 5 | 309 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |