Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011953.1 | NP_001011953.1 | 5 - 6 | 496 | 107 | |
XM_008761549.1 | XP_008759771.1 | 5 - 6 | 522 | 107 | ||
GenBank mRNA | BC083636.1 | - | 5 - 6 | 496 | 7134753 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |