Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105741.1 | NP_001099211.1 | 4 - 5 | 155 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |