Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eepd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014088.3 | NP_001014110.1 | 1 - 2 | 930 | 80 | |
XM_006242726.2 | XP_006242788.1 | 2 - 3 | 1364 | 80 | ||
GenBank mRNA | BC083816.1 | - | 1 - 2 | 930 | 7110630 | 80 |
BC099214.1 | - | 2 - 3 | 1322 | 7384643 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |