Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034150.1 | NP_001029322.1 | 2 - 3 | 309 | 113 | |
GenBank mRNA | BC105782.1 | - | 2 - 3 | 309 | 5599400 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |