Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108247.2 | NP_001101717.1 | 3 - 4 | 492 | 99 | |
XM_006256719.2 | XP_006256781.1 | 4 - 5 | 1033 | 99 | ||
XM_006256720.2 | XP_006256782.1 | 4 - 5 | 1290 | 99 | ||
XM_008773075.1 | XP_008771297.1 | 4 - 5 | 1261 | 99 | ||
GenBank mRNA | BC167040.1 | - | 3 - 4 | 492 | 7135266 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |