Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003976.1 | NP_001003976.1 | 2 - 3 | 408 | 94 | |
XM_006256069.2 | XP_006256131.1 | 2 - 3 | 960 | 94 | ||
XM_008772746.1 | XP_008770968.1 | 2 - 3 | 960 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |