Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 12 - 13 | 1017 | 78 | |
XM_006244296.2 | XP_006244358.1 | 12 - 13 | 1075 | 78 | ||
XM_008766751.1 | XP_008764973.1 | 13 - 14 | 1533 | 78 | ||
GenBank mRNA | AF482705.1 | - | 12 - 13 | 1017 | 78 | |
BC086988.1 | - | 12 - 13 | 1028 | 7192831 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |