Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139336.1 | NP_647552.1 | 8 - 9 | 669 | 120 | |
XM_006244296.2 | XP_006244358.1 | 8 - 9 | 727 | 120 | ||
XM_008766746.1 | XP_008764968.1 | 9 - 10 | 1185 | 120 | ||
XM_008766747.1 | XP_008764969.1 | 837 | 120 | |||
XM_008766748.1 | XP_008764970.1 | 9 - 10 | 1170 | 120 | ||
XM_008766749.1 | XP_008764971.1 | 8 - 9 | 729 | 120 | ||
XM_008766750.1 | XP_008764972.1 | 8 - 9 | 714 | 120 | ||
XM_008766751.1 | XP_008764973.1 | 9 - 10 | 1185 | 120 | ||
XM_008766752.1 | XP_008764974.1 | 8 - 9 | 1077 | 120 | ||
XM_008766753.1 | XP_008764975.1 | 8 - 9 | 1062 | 120 | ||
GenBank mRNA | AF482705.1 | - | 8 - 9 | 669 | 120 | |
BC086988.1 | - | 8 - 9 | 680 | 7192831 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |