Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109064.1 | NP_001102534.1 | 3 - 4 | 835 | 116 | |
XM_006249025.2 | XP_006249087.1 | 3 - 4 | 1067 | 116 | ||
XM_006249026.2 | XP_006249088.1 | 3 - 4 | 853 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |