Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133401.1 | NP_596892.1 | 17 - 18 | 2266 | 164 | |
XM_006235994.2 | XP_006236056.1 | 19 - 20 | 3091 | 164 | ||
GenBank mRNA | AF257746.1 | - | 17 - 18 | 2266 | 164 | |
AF286167.2 | - | 16 - 17 | 2180 | 164 | ||
AY582535.1 | - | 16 - 17 | 2181 | 164 | ||
S66618.1 | - | 4 - 5 | 631 | 164 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |