Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240732.2 | XP_006240794.1 | 5 - 6 | 915 | 80 | |
XM_008765052.1 | XP_008763274.1 | 4 - 5 | 800 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |