Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fopnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108261.1 | NP_001101731.1 | 4 - 5 | 498 | 56 | |
XM_006245848.2 | XP_006245910.1 | 4 - 5 | 504 | 56 | ||
GenBank mRNA | BC083733.1 | - | 4 - 5 | 449 | 56 | |
BC158662.1 | - | 4 - 5 | 464 | 8369763 | 56 | |
FQ215640.1 | - | 4 - 5 | 464 | 56 | ||
FQ228774.1 | - | 4 - 5 | 472 | 56 | ||
FQ228837.1 | - | 5 - 6 | 922 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |