Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108770.1 | NP_001102240.1 | 4 - 5 | 366 | 151 | |
XM_006243332.2 | XP_006243394.1 | 4 - 5 | 350 | 151 | ||
XM_006243337.2 | XP_006243399.1 | 4 - 5 | 389 | 151 | ||
XM_006243341.2 | XP_006243403.1 | 4 - 5 | 389 | 151 | ||
XM_008766315.1 | XP_008764537.1 | 4 - 5 | 610 | 151 | ||
XM_008766316.1 | XP_008764538.1 | 4 - 5 | 463 | 151 | ||
XM_008766317.1 | XP_008764539.1 | 4 - 5 | 350 | 151 | ||
XM_008766318.1 | XP_008764540.1 | 4 - 5 | 350 | 151 | ||
XM_008766319.1 | XP_008764541.1 | 4 - 5 | 389 | 151 | ||
XM_008766320.1 | XP_008764542.1 | 4 - 5 | 350 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |