Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228013.2 | XP_006228075.1 | 1 - 2 | 110 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Transporter Rattus norvegicus Probe spans exons |