Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001717.1 | NP_001001717.1 | 2 | 281 | 66 | |
XM_008772690.1 | XP_008770912.1 | 1 | 389 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |