Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013106.1 | NP_001013124.1 | 6 - 7 | 739 | 87 | |
GenBank mRNA | BC091366.1 | - | 6 - 7 | 739 | 7319183 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |