Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106795.1 | NP_001100265.1 | 4 - 5 | 543 | 113 | |
XM_006242387.2 | XP_006242449.1 | 3 - 4 | 460 | 113 | ||
XM_008765704.1 | XP_008763926.1 | 3 - 4 | 503 | 113 | ||
XM_008765705.1 | XP_008763927.1 | 3 - 4 | 484 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |