Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106453.1 | NP_001099923.1 | 3 - 4 | 1797 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |