Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109655.2 | NP_001103125.1 | 6 - 7 | 566 | 131 | |
XM_006232988.2 | XP_006233050.1 | 7 - 8 | 1111 | 131 | ||
XM_006232989.2 | XP_006233051.1 | 7 - 8 | 1089 | 131 | ||
XM_006232990.2 | XP_006233052.1 | 7 - 8 | 1071 | 131 | ||
GenBank mRNA | EF187253.1 | - | 6 - 7 | 566 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |