Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252167.2 | XP_006252229.1 | 3 - 4 | 922 | 67 | |
XM_006252168.2 | XP_006252230.1 | 2 - 3 | 450 | 67 | ||
XM_006252169.2 | XP_006252231.1 | 3 - 4 | 922 | 67 | ||
XM_008770792.1 | XP_008769014.1 | 3 - 4 | 922 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |