Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scara5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135855.1 | NP_001129327.1 | 2 - 3 | 454 | 105 | |
XM_006252204.2 | XP_006252266.1 | 2 - 3 | 492 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |