Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025142.2 | NP_001020313.2 | 3 - 4 | 808 | 79 | |
GenBank mRNA | BC085942.1 | - | 3 - 4 | 771 | 7126503 | 79 |
BC094955.1 | - | 1 - 2 | 204 | 79 | ||
FQ224662.1 | - | 3 - 4 | 801 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |