Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M83297.1 | - | 3 - 4 | 375 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |