Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022597.2 | NP_072119.2 | 1 - 2 | 110 | 86 | |
GenBank mRNA | BC072490.1 | - | 1 - 2 | 73 | 7097817 | 86 |
CB697852.1 | - | 1 - 2 | 121 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |