Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135756.1 | NP_001129228.1 | 2 - 3 | 393 | 95 | |
XM_008771227.1 | XP_008769449.1 | 1 - 2 | 1047 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |