Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017193.1 | NP_058889.1 | 10 - 11 | 1136 | 63 | |
XM_006253067.2 | XP_006253129.1 | 10 - 11 | 1224 | 63 | ||
GenBank mRNA | BC078864.1 | - | 10 - 11 | 1132 | 7109210 | 63 |
Z50144.1 | - | 10 - 11 | 1136 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |