Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101005.1 | NP_001094475.1 | 5 - 6 | 751 | 147 | |
XM_006235253.2 | XP_006235315.1 | 4 - 5 | 744 | 147 | ||
GenBank mRNA | BC083575.1 | - | 5 - 6 | 751 | 7112877 | 147 |
FQ209572.1 | - | 4 - 5 | 704 | 147 | ||
FQ225908.1 | - | 4 - 5 | 641 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |