Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012962.1 | NP_037094.1 | 10 - 11 | 1077 | 142 | |
XM_008762303.1 | XP_008760525.1 | 10 - 11 | 1491 | 142 | ||
GenBank mRNA | BC078700.1 | - | 9 - 10 | 1042 | 7110971 | 142 |
L38615.1 | - | 10 - 11 | 1077 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |