Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201655.1 | NP_970617.1 | 5 - 6 | 705 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |