Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cx3cl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134455.1 | NP_604450.1 | 2 - 3 | 211 | 90 | |
XM_008772329.1 | XP_008770551.1 | 3 - 4 | 948 | 90 | ||
GenBank mRNA | AF030358.2 | - | 2 - 3 | 211 | 90 | |
BC070938.1 | - | 2 - 3 | 212 | 7099250 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |