Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053392.1 | NP_445844.1 | 7 - 8 | 1479 | 76 | |
XM_006255059.2 | XP_006255121.1 | 7 - 8 | 3559 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |